missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSRP3 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-97433CL1
This item is not returnable.
View return policy
Description
CSRP3 Polyclonal antibody specifically detects CSRP3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CSRP3 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| Cardiac LIM protein, CLPMGC14488, CMD1Mcardiac LIM domain protein, CMH12, CRP3cysteine and glycine-rich protein 3, cysteine and glycine-rich protein 3 (cardiac LIM protein), Cysteine-rich protein 3, LIM domain only 4, LIM domain protein, cardiac, LMO4, MLPMGC61993, Muscle LIM protein | |
| Produced in rabbits immunized with E. coli-derived Human CSRP3 fragment. | |
| 0.1 mL | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| 8048 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction