missing translation for 'onlineSavingsMsg'
Learn More

CSRP3 Antibody (6D2), Novus Biologicals™

Product Code. 18349128 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349128 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349128 Supplier Novus Biologicals Supplier No. H00008048M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CSRP3 Monoclonal antibody specifically detects CSRP3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSRP3
Applications Western Blot, ELISA, Immunocytochemistry, KnockDown
Classification Monoclonal
Clone 6D2
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_003467
Gene Alias Cardiac LIM protein, CLPMGC14488, CMD1Mcardiac LIM domain protein, CMH12, CRP3cysteine and glycine-rich protein 3, cysteine and glycine-rich protein 3 (cardiac LIM protein), Cysteine-rich protein 3, LIM domain only 4, LIM domain protein, cardiac, LMO4, MLPMGC61993, Muscle LIM protein
Host Species Mouse
Immunogen CSRP3 (NP_003467, 95 a.a. ~ 194 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SPKPARSVTTSNPSKFTAKFGESEKCPRCGKSVYAAEKVMGGGKPWHKTCFRCAICGKSLESTNVTDKDGELYCKVCYAKNFGPTGIGFGGLTQQVEKKE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 8048
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.