missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSRP2BP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92912-0.02ml
This item is not returnable.
View return policy
Description
CSRP2BP Polyclonal antibody specifically detects CSRP2BP in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| CSRP2BP | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| ADA2A-containing complex subunit 2, ATAC component 2 homolog, ATAC2CRP2BPKAT14, CRP2 binding partner, CRP2 binding protein, CRP2-binding partner, CSRP2 binding protein, CSRP2-binding protein, cysteine rich protein 2 binding protein, cysteine-rich protein 2-binding protein, dJ717M23.1, MGC15388, PRO1194 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 523-782 of human CSRP2BP (NP_065397.3). DGIYGAKEGGISRLPAGQATYRTTCQDFRILDRYQTSLPSRKGFRHQTTKFLYRLVGSEDMAVDQSIVSPYTSRILKPYIRRDYETKPPKLQLLSQIRSHLHRSDPHWTPEPDAPLDYCYVRPNHIPTINSMCQEFFWPGIDLSECLQYPDFSVVVLYKKVIIAFGFMVPDVKYNEAYISFLFVHPEWRRAGIATFMIYHLIQTCMGKDVTLHVSASNPAMLLYQKFGFKTEEYVLDFYDKYYPLESTECKHAFFLRLRR | |
| 0.02 mL | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 57325 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction