missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSRP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10892-2-AP-150UL
This item is not returnable.
View return policy
Description
CSRP2 is a member of the CSRP family of genes, encoding a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. CRP2 contains two copies of the cysteine-rich amino acid sequence motif (LIM) with putative zinc-binding activity, and may be involved in regulating ordered cell growth. Other genes in the family include CSRP1 and CSRP3.Specifications
| CSRP2 | |
| Polyclonal | |
| Unconjugated | |
| CSRP2 | |
| CRP2, CSRP2, Cysteine rich protein 2, LIM domain only protein 5, LMO 5, LMO5, SmLIM, Smooth muscle cell LIM protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13008, 1466, 29317 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P97314, Q16527, Q62908 | |
| Csrp2 | |
| CSRP2 Fusion Protein Ag1338 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction