missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSRP1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98292CL1
This item is not returnable.
View return policy
Description
CSRP1 Polyclonal antibody specifically detects CSRP1 in Mouse samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
| CSRP1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| CRP, CRP1, CSRPCYRP, cysteine and glycine-rich protein 1, Cysteine-rich protein 1, D1S181E, DKFZp686M148, LIM-domain protein | |
| Produced in rabbits immunized with purified, recombinant Mouse CSRP1 (Uniprot#: P97315; Met1-Glu193) | |
| 0.1 mL | |
| Stem Cells | |
| 1465 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction