missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ CSPD™ Substrate (25 mM Concentrate)

Product Code. 12634315
Click to view available options
Quantity:
0.5 mL
1 mL
2 mL
5 mL
10 mL
25 mL
100 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12634315

Brand: Molecular Probes™ T2041

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This concentrated CSPD™ substrate is a chemiluminescent alkaline phosphatase substrate for protein or nucleic acid blotting on nitrocellulose membranes. This substrate can also be used in solution-based assays.

This CSPD™ chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.

• Chemiluminescent substrates provide highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP).
• Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.

This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours. Solution assays require chemiluminescent enhancers. Membrane-based assays may also require them for increased light output and sensitivity.

For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the product at 4°C.
Form Liquid
For Use With (Application) Southern, Northern, and Western Blotting
Product Line CSPD, NovaBright
Quantity 2 mL
Substrate Alkaline Phosphatase
Substrate Type AP (Alkaline Phosphatase) Substrate
Format Bottle(s)
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.