Learn More
Invitrogen™ CSP alpha Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA1776
Description
PA1-776 detects Cysteine String Protein Alpha in mouse and rat brain samples. PA1-776 has been successfully used in Western blot procedures. By Western blot, this antibody detects a ~34 kDa protein representing Cysteine String Protein Alpha in rat and mouse brain homogenates. The PA1-776 immunogen is a synthetic peptide corresponding to residues E(180) T T Q L T A D S H P S Y H T D G F N(198) of rat CSP. This sequence is conserved across many species including mouse, human and bovine. This peptide (Cat. # PEP-287) is available for use in neutralization and control experiments.
Cysteine string protein alpha (CSPalpha) is a synaptic vesicle protein that contains a DNA-J domain characteristic of Hsp40 chaperones. Cysteine-string proteins are membrane-bound proteins associated primarily with nerve endings and synaptic vesicles in the brain. These proteins play an important role in presynaptic function and may be involved in calcium-dependent neurotransmitter release. CSP proteins may act as presynaptic chaperones that assist in maintaining synaptic function.
Specifications
| CSP alpha | |
| Polyclonal | |
| Unconjugated | |
| Dnajc5 | |
| 2610314I24Rik; AU018536; Ceroid-lipofuscinosis neuronal protein 4; CLN4; CLN4B; Csp; CSP alpha; cysteine string protein; cysteine string protein alpha; DnaJ (Hsp40) homolog, subfamily C, member 5; DnaJ heat shock protein family (Hsp40) member C5; dnaJ homolog subfamily C member 5; Dnajc5; DNAJC5A; mir-941-2; mir-941-3; mir-941-4; mir-941-5; NCL | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13002, 79130, 80331 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| P60904, P60905, Q9H3Z4 | |
| Dnajc5 | |
| Synthetic peptide corresponding to residues E(180) T T Q L T A D S H P S Y H T D G F N(198) of rat CSP. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.