Learn More
CSNK2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20234-1-AP-150UL
 View more versions of this product
Product Code. 16869754
  • £349.00 / 150µL
Description
The antibody is specific to CSNK2B.
This gene encodes the beta subunit of casein kinase II, a ubiquitous protein kinase which regulates metabolic pathways, signal transduction, transcription, translation, and replication. The enzyme is composed of three subunits, alpha, alpha prime and beta, which form a tetrameric holoenzyme. The alpha and alpha prime subunits are catalytic, while the beta subunit serves regulatory functions. The enzyme localizes to the endoplasmic reticulum and the Golgi apparatus.
Specifications
Specifications
CSNK2B
Polyclonal
Unconjugated
CSNK2B
Casein kinase II subunit beta, CK II beta, CK2B, CK2N, CSK2B, CSNK2B, G5A, Phosvitin, Protein G5a
Rabbit
Antigen Affinity Chromatography
RUO
13001, 1460, 81650
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P67870, P67871, P67874
Csnk2b
Peptide
150 μL
Primary
Human, Mouse, Rat