missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSNK2A1 (CK2 α 1) Recombinant Human Protein
CSNK2A1 (CK2 alpha 1)
Brand: Invitrogen PV3248
Code : 50
Additional Details : Weight : 3.36000kg
Description
CSNK2A1 is a serine⁄threonine protein kinase that phosphorylates acidic proteins.Specifications
NP_001883.4 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
47.9kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
CSNK2A1 (CK2 a 1) | |
CSNK2A1 (CK2 alpha 1) was pre-diluted in enzyme dilution buffer and assayed in 50mM HEPES (pH 7.5), 10mM MgCl2, 2.5mM DTT, 0.01% Triton™ X-100, 10% glycerol, 200μM ATP, 100μM casein Kinase 2 peptide substrate (RRRDDDSDDD) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
CSNK2A1 (CK2 α± 1) Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.41mg/mL | |
1457 | |
CSNK2A1 (CK2 α± 1) Recombinant Human Protein | |
–80°C | |
1 Each | |
CSNK2A1 (CK2 α± 1) Recombinant Human Protein | |
CK2 | |
1420nmoles of phosphate transferred to casein kinase 2 peptide substrate (RRRDDDSDDD) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL | |
His Tag | |
Kinase and inhibitors | |
85% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.