missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CSNK1D (CK1 delta), GST Tag Recombinant Protein

Product Code. 15341521
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
This item is not returnable. View return policy

Product Code. 15341521

Brand: Invitrogen™ PR6695B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length

CSNK1D (CK1 delta), a serine/threonine kinase, plays a critical role in Wnt/beta-catenin signaling pathway. This pathway is important in both development and cancer. The encoded protein may also be involved in the regulation of apoptosis, circadian rhythm, microtubule dynamics, chromosome segregation, and p53-mediated effects on growth. The encoded protein is highly similar to the mouse and rat CK1 delta homologs. Three transcript variants encoding different isoforms have been found for CK1 delta.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P48730
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 1453
Molecular Weight (g/mol) 74.3 kDa
Name Human CSNK1D (CK1 delta), GST Tag
Quantity 100 μg
Regulatory Status RUO - research use only
Gene Alias 1200006A05Rik; AA409348; ASPS; casein kinase 1; casein kinase 1 delta; casein kinase 1, delta; casein kinase I isoform delta; CK1d; CK1delta; CKId; CKIdelta; CKI-delta; CSNK1D; D930010H05Rik; FASPS2; HCKID; HGNC:2452; Tau-protein kinase CSNK1D
Common Name CK1 delta
KinaseFamily CK1 Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.