missing translation for 'onlineSavingsMsg'
Learn More

CSNK1A1L Antibody, Novus Biologicals™

Product Code. 18430469 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430469 0.05 mg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430469 Supplier Novus Biologicals Supplier No. H00122011B03P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

CSNK1A1L Polyclonal antibody specifically detects CSNK1A1L in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSNK1A1L
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_660204.1
Gene Alias casein kinase 1, alpha 1-like, casein kinase I alpha S-like, casein kinase I isoform alpha-like, CK1, CKI-alpha-like, EC 2.7.11, EC 2.7.11.1, MGC33182
Host Species Mouse
Immunogen CSNK1A1L (NP_660204.1, 1 a.a. - 337 a.a.) full-length human protein. MTNNSGSKAELVVGGKYKLVRKIGSGSFGDVYLGITTTNGEEVAVKLESQKVKHPQLLYESKLYTILQGGVGIPHMHWYGQEKDNNVLVMDLLGPSLEDLFNFCSRRFTMKTVLMLADQMISRIEYVHTKNFLHRDIKPDNFLMGTGRHCNKLFLIDFGLAKKYRDNRTRQHIPYREDKHLIGTVRYASINAHLGIEQSRRDDMESLGYVFMYFNRTSLPWQGLKAMTKKQKYEKISEKKMSTPVEVLCKGFPAEFAMYLNYCRGLRFEEVPDYMYLRQLFRILFRTLNHQYDYTFDWTMLKQKAAQQAASSSGQGQQAQTQTGKQTEKNKNNVKDN
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 122011
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.