missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSN3 (CASK) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583299
This item is not returnable.
View return policy
Description
Immunogen sequence: NQKQPACHEN DERPFYQKTA PYVPMYYVPN SYPYYGTNLY QRRPAIAINN PYVPRTYYAN Highest antigen sequence indentity to the following orthologs: Rat - 27%, Mouse - 28%.
Kappa-casein (CSN3) stabilizes micelle formation, preventing casein precipitation in milk.
Specifications
| CSN3 (CASK) | |
| Polyclonal | |
| Unconjugated | |
| CSN3 | |
| casein kappa; CASK; CNS10; CSN10; CSN3; CSNK; kappa-casein; KCA | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1448 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P07498 | |
| CSN3 | |
| Recombinant protein corresponding to Human CSN3. Recombinant protein control fragment (Product #RP-105912). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction