missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CSN3 (CASK) Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5122061

 View more versions of this product

Product Code. 17793454

  • £155.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CSN3 (CASK) Polyclonal Antibody for Western Blot, IHC (P)

Kappa-casein (CSN3) stabilizes micelle formation, preventing casein precipitation in milk.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CSN3 (CASK)
Polyclonal
Unconjugated
CSN3
casein kappa; CASK; CNS10; CSN10; CSN3; CSNK; kappa-casein; KCA
Rabbit
Antigen affinity chromatography, Protein A
RUO
1448, 281728
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P02668, P07498
CSN3
Recombinant protein Casein Kappa. The antigen corresponds to amino acid range 24-181 of the target protein.
20 μL
Primary
Bovine, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.