missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSN3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | CSN3 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18284034
|
Novus Biologicals
NBP2-58781 |
100 μL |
£470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18658887
|
Novus Biologicals
NBP2-58781-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CSN3 Polyclonal specifically detects CSN3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CSN3 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| casein kappa, casein, kappa, CASK, CSN10kappa-casein, CSNK, KCA | |
| CSN3 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 1448 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:NQKQPACHENDERPFYQKTAPYVPMYYVPNSYPYYGTNLYQRRPAIAINNPYVPRTYYAN | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title