missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSMD3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-93387
This item is not returnable.
View return policy
Description
CSMD3 Polyclonal specifically detects CSMD3 in Human, Mouse, Rat, Canine samples. It is validated for Peptide ELISA.
Specifications
| CSMD3 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CUB and sushi domain-containing protein 3, CUB and Sushi multiple domains 3, KIAA1894CUB and sushi multiple domains protein 3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 114788 | |
| Human, Mouse, Rat, Canine | |
| IgG |
| ELISA | |
| 0.5 mg/mL | |
| Peptide ELISA Detection limit 1:16000 | |
| NP_937756.1 | |
| CSMD3 | |
| Peptide with sequence KQRTAPKTQYTGC corresponding to internal region according to NP_937756.1, NP_937757.1, NP_443132.2. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to recognize all reported isoforms according to NP_937756.1; NP_937757.1 and NP_443132.2. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction