Learn More
Invitrogen™ CSMD1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA550645
Product Code. 15644575
  • £448.00 / 100µL
Description
The antibody detects endogenous levels of total CSMD1 protein.
CSMD1 CUB and Sushi multiple domains 1 is a protein that in humans is encoded by the CSMD1gene. It is a potential tumour suppressor, the deletion of which may result in head and neck carcinomas behaving more aggressively. CSMD1 is part of the complement system that defends against pathogens through either the classical pathway or the alternative pathway. Located primarily in nerve growth cones, CSMD1 blocks the classical pathway of the immune system and is thought to be involved in tumor suppression, as defects in the gene encoding CSMD1 are associated with squamous cell carcinomas.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CSMD1
Polyclonal
Unconjugated
CSMD1
B930082H09; Csmd1; CUB and sushi domain-containing protein 1; CUB and Sushi multiple domains 1; CUB and sushi multiple domains protein 1; KIAA1890; Mdcr1; mKIAA1890; multiple domain complement regulator 1; PPP1R24; protein phosphatase 1, regulatory subunit 24; UNQ5952/PRO19863
Rabbit
Antigen affinity chromatography
RUO
64478, 94109
-20°C
Liquid
Immunohistochemistry (Paraffin)
3 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q923L3, Q96PZ7
CSMD1
Synthetic peptide corresponding to residues near the C terminal of human CUB and Sushi multiple domains 1.
100 μL
Primary
Human, Mouse