missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSMD1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-29341
This item is not returnable.
View return policy
Description
CSMD1 Polyclonal specifically detects CSMD1 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| CSMD1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CUB and sushi domain-containing protein 1, CUB and sushi multiple domains protein 1, KIAA1890, UNQ5952/PRO19863 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 64478 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:128000 | |
| NP_150094.4 | |
| CSMD1 | |
| Peptide with sequence C-KGVREVNETVTKTP corresponding to internal region according to NP_150094.4. | |
| 0.1 mg | |
| Primary | |
| Reacts with Human. Expected from sequence similarity: Mouse, Rat, Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction