missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CSM-Arg (Powder)
Shop All MP Biomedicals Products
Click to view available options
:
100g
Description
Single Dropout Complete Supplement Mixture used to select for yeast containing plasmids with specific nutritional markers and to select for gene function in transformation experiments. Add 0.74g of CSM-Arg to each liter of purified water and DOB or DOBA. Then, autoclave at 121°C for 25 minutes for complete synthetic defined single drop-out medium for S.cerevisiae.
Specifications
Specifications
| Form | Powder |
| For Use With (Application) | Single Dropout Complete Supplement Mixture used to select for yeast containing plasmids with specific nutritional markers and to select for gene function in transformation experiments. |
| Quantity | 100 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction