missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ csl/RBPJK Antibody (4E12), Novus Biologicals™

Product Code. 18348588 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348588 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18348588 Supplier Novus Biologicals™ Supplier No. H00003516M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

csl/RBPJK Monoclonal antibody specifically detects csl/RBPJK in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen csl/RBPJK
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4E12
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_976029
Gene Alias CBF-1, csl, H-2K binding factor-2, IGKJRB1RBPSUHCBF1, IGKJRBRBP-J kappa, immunoglobulin kappa J region recombination signal binding protein 1, J kappa-recombination signal-binding protein, KBF2, RBP-JK, RBPJKMGC61669, RBP-Jrecombining binding protein suppressor of hairless (Drosophila), recombination signal binding protein for immunoglobulin kappa J region, recombining binding protein suppressor of hairless, Renal carcinoma antigen NY-REN-30, SUH, suppressor of hairless homolog
Host Species Mouse
Immunogen RBPJ (NP_976029, 3 a.a. ∽ 109 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. WIKRKFGERPPPKRLTREAMRNYLKERGDQTVLILHAKVAQKSYGNEKRFFCPPPCVYLMGSGWKKKKEQMERDGCSEQESQPCAFIGIGNSDQEMQQLNLEGKNYC
Purification Method IgG purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cell Biology, Neuroscience, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 3516
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.