Learn More
Eurofins DiscoverX CSK Protein, Active
Description
CSK (C-SRC, c-src tyrosine kinase) is a non-receptor tyrosine kinase that catalyzes the tyrosine phosphorylation of Src family members at C-terminal, promoting an intramolecular association between the phosphotyrosine and the Src homology 2 (SH2) domain and negatively regulating Src catalytic activity in vivo. Although CSK has substantial homology to Src kinases, it lacks the C-terminal regulatory tyrosine found in Src kinases.
GenBank X60114 (CDS GB 30314)
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl, pH 7.5, 0.1 mM EGTA, 150 mM NaCl, 1 mM benzamidine, 0.03% Brij-35, 0.2 mM PMSF, 0.1% 2-mercaptoethanol, 50% glycerol. Liquid at -20°C |
| Molecular Weight (g/mol) | 77.6 kDa |
| Quantity | 250 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal GST incorporating PreScission Protease cleavage site |
| Expression System | Recombinant enzyme expressed in E. coli cells |
| Protein Form | Full-length |
| Purity or Quality Grade | ≥75% |
| Protein | CSK, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.