missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This is one of the several different receptors for 5-hydroxytryptamine, a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that stimulate adenylate cyclase. It has a high affinity for tricyclic psychotropic drugs.
Specifications
Specifications
| Antigen | CSK |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 3B12C5 |
| Concentration | 0.89 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CSK |
| Gene Accession No. | P41240, P41241 |
| Gene Alias | C Src kinase, c src tyrosine kinase, CSK, Protein tyrosine kinase CYL, Tyrosine protein kinase CSK |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?