missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSK Binding Protein Antibody (PAG-C1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-42418
This item is not returnable.
View return policy
Description
CSK Binding Protein Monoclonal antibody specifically detects CSK Binding Protein in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin).
Specifications
| CSK Binding Protein | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin | |
| C-terminal peptide (last 15 amino acids) of human Csk binding protein coupled to KLH. | |
| 0.05 mg | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| PAG-C1 | |
| Unconjugated | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| The antibody recognizes an epitope located in the C-terminal domain of Csk-binding protein (Cbp), also known as protein associated with glycosphingolipid-enriched microdomains (PAG). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction