missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSK Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-808
This item is not returnable.
View return policy
Description
CSK Polyclonal specifically detects CSK in Human, Mouse, Rat samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| CSK | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| C-Src kinase, c-src tyrosine kinase, EC 2.7.10, EC 2.7.10.2, MGC117393, Protein-tyrosine kinase CYL, tyrosine-protein kinase CSK | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Reported variants represent identical protein: NP_004374.1; NP_001120662.1. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 ug/mL, Peptide ELISA Detection limit 1:32000 | |
| NP_004374.1 | |
| CSK | |
| Peptide with sequence C-EQLEHIKTHELHL corresponding to C-Terminus according to NP_004374.1, NP_001120662.1. | |
| 0.1 mg | |
| Cell Cycle and Replication, Signal Transduction | |
| 1445 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction