missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSHL1 Recombinant Rabbit Monoclonal Antibody (K01_1P85)
Description
CSHL1 Recombinant Monoclonal Antibody for Western Blot
May be a novel gestational hormone required to compensate for absence of other members of the GH/CS cluster during gestation.
Specifications
Specifications
| Antigen | CSHL1 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_1P85 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4 |
| Gene | CSHL1 |
| Gene Accession No. | Q14406 |
| Gene Alias | chorionic somatomammotropin CS-5; chorionic somatomammotropin hormone like 1; chorionic somatomammotropin hormone-like 1; Chorionic somatomammotropin-like; CS-5; CSHL1; CSHP1; CSL; GHB4; growth hormone B4; growth hormone cluster; hCS-L; Lactogen-like |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?