missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSGLCAT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101521
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CSGLCAT.
CSGlcA-T belongs to the chondroitin N-acetylgalactosaminyltransferase family. It transfers glucuronic acid (GlcUA) from UDP-GlcUA to N-acetylgalactosamine residues on the non-reducing end of the elongating chondroitin polymer. CSGlcA-T has no N-acetylgalactosaminyltransferase activity.
Specifications
| CSGLCAT | |
| Polyclonal | |
| Unconjugated | |
| CHPF2 | |
| Chondroitin glucuronyltransferase; Chondroitin polymerizing factor 2; chondroitin sulfate glucuronyltransferase; Chondroitin synthase 3; chondroitin synthase-3; CHPF2; chPF-2; CHSY3; ChSy-3; CSGlcAT; CSGlcA-T; KIAA1402; N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase; UNQ299/PRO339 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 54480 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9P2E5 | |
| CHPF2 | |
| A synthesized peptide derived from human CHPF2(Accession Q9P2E5), corresponding to amino acid residues G36-V86. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction