missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CSGALNACT2 Polyclonal antibody specifically detects CSGALNACT2 in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | CSGALNACT2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS (pH 7.4) |
| Gene Accession No. | NP_061060.3 |
| Gene Alias | beta4GalNAcT-2, CHGN2, chondroitin beta1,4 N-acetylgalactosaminyltransferase 2, Chondroitin beta-1,4-N-acetylgalactosaminyltransferase 2, chondroitin sulfate GalNAcT-2, chondroitin sulfate N-acetylgalactosaminyltransferase 2, DKFZp686H13226, EC 2.4.1.174, FLJ43310, GALNACT-2, GALNACT2beta 4 GalNAcT-2, MGC40204, PRO0082 |
| Host Species | Rabbit |
| Immunogen | CSGALNACT2 (NP_061060.3, 1 a.a. - 542 a.a.) full-length human protein. MPRRGLILHTRTHWLLLGLALLCSLVLFMYLLECAPQTDGNASLPGVVGENYGKEYYQALLQEQEEHYQTRATSLKRQIAQLKQELQEMSEKMRSLQERRNVGANGIGYQSNKEQAPSDLLEFLHSQIDKAEVSIGAKLPSEYGVIPFESFTLMKVFQLEMGLTRHPEEKPVRKDKRDELVEVIEAGLEVINNPDEDDEQEDEEGPLGEKLIFNENDFVEGYYRTERDKGTQYELFFKKADLTEYRHVTLFRPFGPLMKVKSEMIDITRSIINIIVPLAERTEAFVQFMQNFRDVCIHQDKKIHLTVVYFGKEGLSKVKSILESVTSESNFHNYTLVSLNEEFNRGRGLNVGARAWDKGEVLMFFCDVDIYFSAEFLNSCRLNAEPGKKVFYPVVFSLYNPAIVYANQEVPPPVEQQLVHKKDSGFWRDFGFGMTCQYRSDFLTIGGFDMEVKGWGGEDVHLYRKYLHGDLIVIRTPVPGLFHLWHEKRCADELTPEQYRMCIQSKAMNEASHSHLGMLVFREEIETHLHKQAYRTNSEAVG |
| Purification Method | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?