missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSF3R Recombinant Rabbit Monoclonal Antibody

Description
CSF3R Recombinant Monoclonal Antibody for IHC (P)
CSF3R is the receptor for colony stimulating factor 3, a cytokine that controls the production, differentiation, and function of granulocytes. The encoded protein, which is a member of the family of cytokine receptors, may also function in some cell surface adhesion or recognition processes. Four transcript variants encoding four different isoforms have been found for this gene, with three of the isoforms being membrane-bound and the other being secreted and soluble.
Specifications
Specifications
| Antigen | CSF3R |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Recombinant Monoclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide |
| Gene | CSF3R |
| Gene Accession No. | Q99062 |
| Gene Alias | CD114; CD114 antigen; colony stimulating factor 3 receptor; colony stimulating factor 3 receptor (granulocyte); Csf3r; Csfgr; G-CSF receptor; GCSFR; G-CSFR; G-CSF-R; Granulocyte colony-stimulating factor receptor; SCN7 |
| Gene Symbols | CSF3R |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?