Learn More
Sino Biological™ CSF3R (CD114) Recombinant Human Protein, His Tag
Expressed from DNA sequence encoding extracellular domain (Met 1-Pro 621) of human CSF3R precursor fused topolyhistidine tag at C-terminus
Brand: Sino Biological™ 10218H08H5
This item is not returnable.
View return policy
Description
Expressed from DNA sequence encoding extracellular domain (Met 1-Pro 621) of human CSF3R precursor fused topolyhistidine tag at C-terminus
- Measured by its ability to inhibit the CSF3 induced proliferation of NFS60 mouse myeloid cells. The ED50 for this effect is typically 20-80 ng/ml.
Specifications
1441 | |
5μg | |
−20° to −70°C | |
<1 EU/μg | |
CSF3R |
68kd | |
Human | |
Truncated, Cell Surface Protein, Recombinant | |
Colony stimulating factor 3 receptor | |
CSF3R (CD114) Recombinant Human Protein |
For Research Use Only. Not for use in diagnostic procedures.