Learn More
CSF3 Mouse anti-Human, Clone: SPM468, Abnova™
Mouse monoclonal antibody raised against native human CSF3.
Brand: Abnova MAB13047.100ug
Additional Details : Weight : 0.02000kg
Description
The protein encoded by this gene is a cytokine that controls the production, differentiation, and function of granulocytes. The active protein is found extracellularly. Three transcript variants encoding three different isoforms have been found for this gene. [provided by RefSeq]
Specifications
CSF3 | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
G-CSF/GCSF/MGC45931 | |
Mouse | |
Protein A/G purification | |
RUO | |
1440 | |
Human | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
SPM468 | |
Mouse monoclonal antibody raised against native human CSF3. | |
In 10mM PBS. | |
CSF3 | |
Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes. | |
100 μg | |
Primary | |
This antibody recognizes CSF3 in the cytoplasm of mature granulocytes. It shows no reactivity with any other cell types. | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |