missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CSF3 (Human) Recombinant Protein
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µg
50µg
Specifications
Specifications
| Gene ID (Entrez) | 1440 |
| Name | colony stimulating factor 3 (granulocyte) |
| Preparation Method | Hordeum vulgare (barley) expression system |
| Quality Control Testing | SDS-PAGE Stained with Coomassie Blue |
| Quantity | 50 μg |
| Storage Requirements | Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Gene Alias | G-CSF, GCSF, MGC45931 |
| Gene Symbol | CSF3 |
| Cross Reactivity | Human |
| Species | Plants |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction