missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSE1L/CAS/Exportin-2 Antibody (5111), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-43609
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
CSE1L/CAS/Exportin-2 Monoclonal specifically detects CSE1L/CAS/Exportin-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| CSE1L/CAS/Exportin-2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| CSE1L | |
| Recombinant protein encompassing a sequence within the center region of human CSE1L. The exact sequence is proprietary. | |
| 0.1 mL | |
| Apoptosis, Cancer, Core ESC Like Genes, Stem Cell Markers | |
| 1434 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot | |
| 5111 | |
| Western Blot 1:500-1:3000 | |
| CASMGC117283, Cellular apoptosis susceptibility protein, chromosome segregation 1 (yeast homolog)-like, Chromosome segregation 1-like protein, CSE1, CSE1 chromosome segregation 1-like (yeast), CSE1L, Exp2, exportin-2, Importin-alpha re-exporter, MGC130037, XPO2MGC130036 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction