missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSDE1 Recombinant Rabbit Monoclonal Antibody (23GB3510), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553423
This item is not returnable.
View return policy
Description
CSDE1 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
Specifications
| CSDE1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CSDE1 | |
| AA960392; BC016898; cold shock domain containing E1; cold shock domain containing E1, RNA binding; cold shock domain containing E1, RNA-binding; cold shock domain-containing protein E1; CSDE1; D1S155E; D3Jfr1; KIAA0885; mKIAA0885; N-ras upstream gene protein; NRAS-related; NRU; Protein UNR; RP5-1000E10.3; UNR; upstream of NRAS | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 117180, 229663, 7812 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3510 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75534, P18395, Q91W50 | |
| CSDE1 | |
| A synthetic peptide of human CSDE1 (401-450AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction