missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSDE1 Recombinant Rabbit Monoclonal Antibody (23GB3505), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | CSDE1 |
|---|---|
| Clone | 23GB3505 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088951
![]() |
Invitrogen™
MA553420 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088937
|
Invitrogen™
MA553421 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CSDE1 Recombinant Monoclonal antibody specifically detects CSDE1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| CSDE1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O75534, P18395, Q91W50 | |
| 117180, 229663, 7812 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3505 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AA960392; BC016898; cold shock domain containing E1; cold shock domain containing E1, RNA binding; cold shock domain containing E1, RNA-binding; cold shock domain-containing protein E1; CSDE1; D1S155E; D3Jfr1; KIAA0885; mKIAA0885; N-ras upstream gene protein; NRAS-related; NRU; Protein UNR; RP5-1000E10.3; UNR; upstream of NRAS | |
| A synthetic peptide of human CSDE1 (50-100AA). | |
| Primary | |
| -20°C | |
| CSDE1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title