missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSDE1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13319-1-AP-20UL
This item is not returnable.
View return policy
Description
CSDE1 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.Specifications
| CSDE1 | |
| Polyclonal | |
| Unconjugated | |
| CSDE1 | |
| CSDE1, D1S155E, KIAA0885, N ras upstream gene protein, NRU, Protein UNR, RP5 1000E10.3, UNR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 229663, 7812 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75534, Q91W50 | |
| CSDE1 | |
| CSDE1 Fusion Protein Ag4173 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction