Learn More
Invitrogen™ CSDE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596480
Description
Immunogen sequence: LSAQRNHAIR IKKLPKGTVS FHSHSDHRFL GTVEKEATFS NPKTTSPNKG KEKEAEDGII AYDDCGVKLT IAFQAKDVEG STSPQIGDKV EFSISDKQRP GQQVATCVRL LGRNSNSKRL LGYVATLKDN FGFIETANHD KEIFFHYSEF SGDVDSLELG DMVEYSLSKG KGNKVSAEKV NKTHSVNGIT EEADPTIYSG KVIRPLRSVD PTQTEYQGMI EIVEEGDMKG EV; Positive Samples: U-87MG, LO2, HeLa, MCF7, Mouse brain; Cellular Location: Cytoplasm.
CSDE1 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
Specifications
| CSDE1 | |
| Polyclonal | |
| Unconjugated | |
| CSDE1 | |
| AA960392; BC016898; cold shock domain containing E1; cold shock domain containing E1, RNA binding; cold shock domain containing E1, RNA-binding; cold shock domain-containing protein E1; CSDE1; D1S155E; D3Jfr1; KIAA0885; mKIAA0885; N-ras upstream gene protein; NRAS-related; NRU; Protein UNR; RP5-1000E10.3; UNR; upstream of NRAS | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 229663, 7812 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.74 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75534, Q91W50 | |
| CSDE1 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.