missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CSDE1 Antibody (SR4307), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27842-100ul
This item is not returnable.
View return policy
Description
CSDE1 Monoclonal antibody specifically detects CSDE1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| CSDE1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7812 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| SR4307 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| cold shock domain containing E1, RNA-binding, cold shock domain-containing protein E1, D1S155EUNRRP5-1000E10.3, DKFZp779B0247, DKFZp779J1455, FLJ26882, KIAA0885, N-ras upstream gene protein, NRAS-related, NRU, Protein UNR, upstream of NRAS | |
| A synthesized peptide derived from human CSDE1 (Uniprot #: O75534) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction