missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSDC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551815
This item is not returnable.
View return policy
Description
Immunogen sequence: PTFPFHREGS RVWERGGVPP RDLPSPLPTK RTRTYSATAR ASAGPVFKGV CKQFSRSQGH GFITPENGSE DIFVHVSDIE GEYVPVEGDE VTYKMCPIPP KNQ Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 96%.
RNA-binding factor which binds specifically to the very 3'-UTR ends of both histone H1 and H3.3 mRNAs, encompassing the polyadenylation signal. Might play a central role in the negative regulation of histone variant synthesis in the developing brain.
Specifications
| CSDC2 | |
| Polyclonal | |
| Unconjugated | |
| CSDC2 | |
| AI415250; AI481750; cold shock domain containing C2; cold shock domain containing C2, RNA binding; cold shock domain-containing protein C2; CSDC2; dJ347H13.2; PIPPIN; RNA-binding protein pippin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27254 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y534 | |
| CSDC2 | |
| Recombinant protein corresponding to Human CSDC2. Recombinant protein control fragment (Product #RP-91521). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction