missing translation for 'onlineSavingsMsg'
Learn More

CSAP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£169.00 - £255.00

Specifications

Antigen CSAP
Dilution Immunohistochemistry-Paraffin 1:200-1:1000
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18633552
View Documents
Novus Biologicals
NBP2-98036-100ul
100 μg
£255.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18670704
View Documents
Novus Biologicals
NBP2-98036-50ul
50 μg
£169.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CSAP Polyclonal antibody specifically detects CSAP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

CSAP
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
C1orf96, centriole, cilia and spindle-associated protein, chromosome 1 open reading frame 96, RP4-803J11.3
Produced in rabbits immunized with E. coli-derived Human CSAP fragment.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunohistochemistry-Paraffin 1:200-1:1000
Polyclonal
Purified
RUO
PBS (pH 7.0)
126731
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.