Learn More
BD Biosciences™ CS&T IVD Beads (CE/IVD)
Click to view available options
No. of Tests:
150 Tests
50 Tests
Tamaño de la unidad:
Each
Descripción
- Intended for in vitro diagnostic use on BD FACSCanto™ II flow cytometers running BD FACSDiva™ software.
- Used to set up the cytometer, to perform daily performance quality control (QC), and to determine lyse/wash (LW) application settings.
- Dyed with fluorochromes that are excited by the cytometer's lasers.
Especificaciones
Especificaciones
| Content And Storage | Store vials at 2°C–8°C and protect from light. |
| No. of Tests | 50 Tests |
| Stability | The beads are stable until the date shown on the vial label when stored as directed. Do not use after the expiration date. After dilution, the beads are stable for 8 hours at 2°C–25°CC |
| Product Type | Fluorescent Polystyrene Beads |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido