Learn More
CS Citrate Synthase Antibody (CL2579) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-36775AF532
Product Code. 30290555
  • £558.00 / 0.10mL
Description
CS Citrate Synthase Monoclonal antibody specifically detects CS Citrate Synthase in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CS Citrate Synthase
Monoclonal
Alexa Fluor 532
citrate synthase, citrate synthase, mitochondrial, EC 2.3.3, EC 2.3.3.1
This antibody was developed using a recombinant protein derived from O75390, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Endocrinology, Signal Transduction
1431
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL2579
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified