missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CS Citrate Synthase Antibody (CL2548) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-36772AF700
This item is not returnable.
View return policy
Beskrivning
CS Citrate Synthase Monoclonal antibody specifically detects CS Citrate Synthase in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifikationer
| CS Citrate Synthase | |
| Monoclonal | |
| Alexa Fluor 700 | |
| citrate synthase, citrate synthase, mitochondrial, EC 2.3.3, EC 2.3.3.1 | |
| This antibody was developed using a recombinant protein derived from O75390, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 1431 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL2548 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering