Learn More
CS Citrate Synthase Antibody (1761), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-43648
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry (Paraffin)
CS Citrate Synthase Monoclonal specifically detects CS Citrate Synthase in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| citrate synthase, citrate synthase, mitochondrial, EC 2.3.3, EC 2.3.3.1 | |
| 52 kDa |