missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRYZ Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582780
This item is not returnable.
View return policy
Description
Immunogen sequence: HGGRVIVVGS RGTIEINPRD TMAKESSIIG VTLFSSTKEE FQQYAAALQA GMEIGWLKPV IGSQYPLEKV AEAHENIIHG SGA Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 80%.
Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. The former class is also called phylogenetically-restricted crystallins. This gene encodes a taxon-specific crystallin protein which has NADPH-dependent quinone reductase activity distinct from other known quinone reductases. It lacks alcohol dehydrogenase activity although by similarity it is considered a member of the zinc-containing alcohol dehydrogenase family. Unlike other mammalian species, in humans, lens expression is low. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. One pseudogene is known to exist.
Specifications
| CRYZ | |
| Polyclonal | |
| Unconjugated | |
| Cryz | |
| crystallin zeta; crystallin, zeta; crystallin, zeta (quinone reductase); Cryz; nadph quinone reductase; NADPH:quinone reductase; quinone oxidoreductase; quinone reductase; Sez9; zeta crystallin; Zeta-crystallin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1429 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q08257 | |
| Cryz | |
| Recombinant protein corresponding to Human CRYZ. Recombinant protein control fragment (Product #RP-93140). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction