missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRYZ Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97243AF647
This item is not returnable.
View return policy
Description
CRYZ Polyclonal antibody specifically detects CRYZ in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CRYZ | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 647 | |
| crystallin, zeta (quinone reductase), DKFZp779E0834, EC 1.6.5.5, FLJ41475, NADPH:quinone reductase, quinone oxidoreductase, Zeta-crystallin | |
| Produced in rabbits immunized with E. coli-derived Human CRYZ fragment. | |
| 0.1 mL | |
| Vision | |
| 1429 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction