missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Crystallized Bovine Albumin
Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 g
10 g
100 g
Unit Size:
100g
10g
5g
Description
- Source: Bovine blood
- Assay (electrophoresis): ≥99%
- Identity: By IR
- Protein: ≥98%
- pH (7%, 0.15M NaCl): 5.0 - 5.4
- Moisture (KF) when packaged: %le;5.0%
- Sodium (Na): 0 - 5.0mg/g
- Chloride (Cl): 0 - 3.0mg/g
- CAS: 9048-46-8
Specifications
Specifications
| pH Range | 5.0 to 5.4 |
| Quantity | 5 g |
| Source | Bovine blood |
| Packaging | Plastic Bottle |
| Product Type | Bovine Albumin |
| Form | Crystalline |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction