Learn More
Crystal Violet Solution (non-aqueous indicator) According to European Pharmacopoeia (EP), Reagecon
High-quality indicator solution produced from the highest quality raw materials and manufactured and tested under rigorous conditions.
Brand: Reagecon 1022901
Code : A0
Additional Details : CAS Number : 64-19-7 Weight : 0.10497kg Transport : UN number : 2920 Chem class : 8 Pack group : II
Description
- European Pharmacopoeia (EP) Reference Standard, European Pharmacopoeia (EP) Reference Material
- High quality indicator solution produced from the highest quality raw materials and manufactured and tested under rigorous conditions.
- For manual titrations; indicator titrant or analyte change color and this color change is by far the most important method of end point detection, in such titrations.
- A high-quality indicator solution such as Reagecon's facilitates end point detection, in a clear and unambiguous manner and is an imperative for accurate manual titrimetry.
Specifications
Amber Glass Winchester Bottle | |
100 mL | |
Liquid |
Safety and Handling
- CRYSTAL VIOLET SOLUTION
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : acetic acid, C.I. Basic Violet 3