missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Crystal Violet Solution
Description
- Pro-Lab Diagnostics offers an extensive range of ready-to-use and concentrated microbiological staining solutions
- Solutions come in a selection of convenient and economical sizes.
- All stains are manufactured in accordance with all current regulatory requirements
- CE labelled
Specifications
Specifications
| Solution Type | Reagent |
| Type | Crystal Violet |
| Quantity | 1L |
| Certifications/Compliance | (EC) No 1907/2006, Annex II, (EU) No 453/2010, CE-mark |
| Format | Liquid |
| For Use With (Application) | For Gram staining |
| Color | Dark violet |
| Composition | Ethanol, C.I. basic violet 3, Methanol |
| Storage Requirements | <20°C |
| Stain Type | Gram Stain |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?