missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Crystal violet, ACS reagent

Product Code. 11986931 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11986931 250 g Glass bottle 250g
11976931 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11986931 Supplier Thermo Scientific Chemicals Supplier No. 447572500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 548-62-9 | C25H30ClN3 | 407.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Crystal violet
CAS 548-62-9, 90-94-8
Melting Point 173°C
Packaging Glass bottle
Molecular Formula C25H30ClN3
Quantity 250 g
Merck Index 15,443
Synonym crystal violet, gentian violet, basic violet 3, methylrosaniline chloride, aniline violet, hexamethyl violet, gentiaverm, hexamethylpararosaniline chloride, pyoktanin, adergon
Solubility Information Solubility in water: 16g/L (25°C). Other solubilities: soluble in chloroform,1g/10 mL alcohol,1g/15 mL glycerin,practically insoluble in ether
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
Molecular Weight (g/mol) 407.99
PubChem CID 11057
ChEBI CHEBI:41688
Formula Weight 407.99
Grade ACS Reagent
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Crystal Violet
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: C.I. Basic Violet 3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.