missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Crystal Violet, ACS, 90+%, Thermo Scientific Chemicals
73.80 GBP valid until 2024-04-26
Use promo code "21687" to get your promotional price.
Use promo code "21687" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
Crystal Violet is used as an acid-base indicator, an alcohol denaturant, a biological stain, a textile dye and an indicator for copper salts. It is a component of gram staining in microorganisms and cell lines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | ∼215°C (decomposition) |
UN Number | UN3077 |
Beilstein | 4077708 |
Merck Index | 14,4395 |
Solubility Information | Soluble in water and chloroform. Slightly soluble in alcohol and glycerol. |
Formula Weight | 407.99 |
Percent Purity | ≥90% |
Grade | ACS |
Quantity | 100 g |
Physical Form | Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'