missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Crystal Violet

Product Code. p-7026117
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435027 25 g 25g
11445027 100 g 100g
11455027 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435027 Supplier Thermo Scientific Chemicals Supplier No. B21932.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol

Crystal Violet is used as an acid-base indicator, alcohol denaturant, biological stain, textile dye, and an indicator for copper salts. It is a component of gram staining in microorganisms and cell lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Crystal Violet is used as an acid-base indicator, alcohol denaturant, biological stain, textile dye, and an indicator for copper salts. It is a component of gram staining in microorganisms and cell lines.

Solubility
Miscible with water and chloroform. Partially miscible with alcohol and glycerol.

Notes
Incompatible with acids, oxidizing agents, alkali metals, strong oxidizing agents, ammonia, acid chlorides, acid anhydrides, reducing agents and peroxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 548-62-9
Molecular Formula C25H30ClN3
Molecular Weight (g/mol) 407.986
MDL Number MFCD00011750
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
Synonym Basic Violet 3; C.I. 42555
PubChem CID 11057
ChEBI CHEBI:41688
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]

Specifications

Melting Point ∼215°C (decomposition)
UN Number UN3077
Beilstein 4077708
Merck Index 14,4395
Solubility Information Miscible with water and chloroform. Partially miscible with alcohol and glycerol.
Formula Weight 407.99
Quantity 25 g
Chemical Name or Material Crystal Violet
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Crystal Violet
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: C.I. Basic violet 1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.